Phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid

B0857-250G B0857-25G B0857-5G
TCB0857-250GEA 425.02 CAD
TCB0857-250G TCB0857-025G TCB0857-005G
Phenylboronic acid (contains varying amounts of Anhydride)
Phenylboronic acid

CAS Number: 98-80-6
MDL Number: MFCD00002103
Molecular Formula: C6H7BO2
Molecular Weight: 121.93
Form: Crystal
Color: White
Melting point (°C): 216


Other Product Names / Synonyms: Benzeneboronic Acid

Formula: C₆H₇BO₂
Melting Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00002103
CAS Number: 98-80-6
EINECS: 202-701-9
Merck Index: 13,01068
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=734d53715068835d4d746ed2356dc5838b77763b&vid=734d53715068835d4d746ed2356dc5838b77763b&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR