4,4'-Dimethyl-2,2'-bipyridyl ≥98.0%

Supplier: TCI America

Synonyms: 4,4'-Dimethyl-2,2'-bipyridine

B0852-1G B0852-5G B0852-25G
TCB0852-001GEA 45.82 CAD
TCB0852-001G TCB0852-005G TCB0852-25G
4,4'-Dimethyl-2,2'-bipyridyl ≥98.0%
4,4'-Dimethyl-2,2'-bipyridyl

CAS Number: 1134-35-6
MDL Number: MFCD00006441
Molecular Formula: C12H12N2
Molecular Weight: 184.24
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 172


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: 2,2'-Bi-4-picoline, 2,2'-Di-4-picolyl

Formula: C₁₂H₁₂N₂
MW: 184.24 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00006441
CAS Number: 1134-35-6
EINECS: 214-483-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b995d2da19f70ab8d6fb4ba94c1a4951339eede&vid=2b995d2da19f70ab8d6fb4ba94c1a4951339eede&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR