9-Bromophenanthrene ≥98.0%

Supplier: TCI America

B0838-250G B0838-25G
TCB0838-250GEA 1033 CAD
TCB0838-250G TCB0838-025G
9-Bromophenanthrene ≥98.0%
9-Bromophenanthrene

CAS Number: 573-17-1
MDL Number: MFCD00001174
Molecular Formula: C14H9Br
Molecular Weight: 257.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 225
Melting point (°C): 63
Flash Point (°C): 110

Formula: C₁₄H₉Br
Boiling Pt: 225 °C
Melting Pt: 63 °C
Density: 1.4093 g/cm³ (101 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001174
CAS Number: 573-17-1
EINECS: 209-351-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fb7d5b547741ccc6bea0eb90d89881116bd306d&vid=1fb7d5b547741ccc6bea0eb90d89881116bd306d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR