2-Bromobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

B0836-500G B0836-25G B0836-100G
TCB0836-500GEA 390.26 CAD
TCB0836-500G TCB0836-025G TCB0836-100G
2-Bromobenzaldehyde ≥98.0%
2-Bromobenzaldehyde

CAS Number: 6630-33-7
MDL Number: MFCD00003300
Molecular Formula: C7H5BrO
Molecular Weight: 185.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Flash Point (°C): 95
Freezing point (°C): 16
Specific Gravity (20/20): 1.60

Formula: C₇H₅BrO
Boiling Pt: 230 °C
Melting Pt: 21 °C
Density: 1.60 (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00003300
CAS Number: 6630-33-7
EINECS: 229-622-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b58b4e3b342b5db44e6e89da867a9abf14c3237&vid=502418375331fbf00d09a59d261b6b0effa48609&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR