1,4-Butanedithiol ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B0829-25ML B0829-5ML
TCB0829-25MLEA 168.59 CAD
TCB0829-25ML TCB0829-5ML
1,4-Butanedithiol ≥95.0% (by GC, titration analysis)
1,4-Butanedithiol

CAS Number: 1191-08-8
MDL Number: MFCD00004906
Molecular Formula: C4H10S2
Molecular Weight: 122.24
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 195
Flash Point (°C): 70
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,4-Dimercaptobutane

Formula: C₄H₁₀S₂
Boiling Pt: 195 °C
Melting Pt: –53 °C
Density: 1.05 (20 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00004906
CAS Number: 1191-08-8
EINECS: 214-728-3
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce6afcfc36feee3056f714a257de326c01b38dab&vid=a4d903dd46ee89bfb01ec14de6f97ae48010e15c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR