Bis(2-butoxyethyl)ether ≥98.0%
Supplier: TCI America
Synonyms:
Diethylene glycol dibutyl ether, 2,2-Dibutoxyethyl ether
B0828-500ML
B0828-25ML
TCB0828-500MLEA
183.28
CAD
TCB0828-500ML
TCB0828-025ML
Bis(2-butoxyethyl)ether ≥98.0%
Bis(2-butoxyethyl) ether
CAS Number: 112-73-2
MDL Number: MFCD00009459
Molecular Formula: C12H26O3
Molecular Weight: 218.34
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Melting point (°C): -60
Flash Point (°C): 122
Specific Gravity (20/20): 0.88
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Bis(2-butoxyethyl) Ether, Dibutyl Carbitol
Formula:
C₁₂H₂₆O₃ MW: 218.34 g/mol Boiling Pt: 255 °C Melting Pt: –60 °C Density: 0.88 (20 °C) Flash Pt: 122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009459 CAS Number: 112-73-2 EINECS: 204-001-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...