2,2'-Dihydroxybiphenyl ≥98.0%
Supplier: TCI America
Synonyms:
2,2'-Biphenol, 2,2-Biphenol, Biphenyl-2,2'-diol
CAS Number: 1806-29-7
MDL Number: MFCD00002210
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 325
Melting point (°C): 107
Flash Point (°C): 154
Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research), Pressure & Heat Sensitive Recording Materials
Other Product Names / Synonyms: Biphenyl-2,2'-diol, 2,2'-Biphenol
Formula:
C₁₂H₁₀O₂ Boiling Pt: 325 °C Melting Pt: 107 °C Density: 1.342 g/cm³ (20 °C) Flash Pt: 154 °C |
MDL Number:
MFCD00002210 CAS Number: 1806-29-7 EINECS: 217-303-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...