2-Bromo-p-cresol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Bromo-4-methylphenol, 1-Bromo-2-hydroxy-5-methylbenzene

B0808-500G B0808-25G
TCB0808-500GEA 1793.3 CAD
TCB0808-500G TCB0808-025G
2-Bromo-p-cresol ≥98.0%
2-Bromo-p-cresol

CAS Number: 6627-55-0
MDL Number: MFCD00002151
Molecular Formula: C7H7BrO
Molecular Weight: 187.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 110
Freezing point (°C): 16
Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: 2-Bromo-4-methylphenol

Formula: C₇H₇BrO
Boiling Pt: 214 °C
Melting Pt: 26 °C
Density: 1.53 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002151
CAS Number: 6627-55-0
EINECS: 229-595-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7ea286485e37219bec8a2114a5306653d1e36dc&vid=d7ea286485e37219bec8a2114a5306653d1e36dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR