(±)-Ethyl-2-bromobutyrate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 2-bromobutyrate, Ethyl 2-bromobutanoate

B0800-500G B0800-25G
TCB0800-500GEA 251.22 CAD
TCB0800-500G TCB0800-25G
(±)-Ethyl-2-bromobutyrate ≥98.0%
(±)-Ethyl 2-bromobutyrate

CAS Number: 533-68-6
MDL Number: MFCD00000155
Molecular Formula: C6H11BrO2
Molecular Weight: 195.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 73
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: 2-Bromobutyric Acid Ethyl Ester

Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 178 °C
Density: 1.33 (20 °C)
Flash Pt: 73 °C
MDL Number: MFCD00000155
CAS Number: 533-68-6
EINECS: 208-574-6
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65f7b77f25c8402e0fffd6ea943ddd397d3d5110&vid=65f7b77f25c8402e0fffd6ea943ddd397d3d5110&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR