Butyrophenone ≥99.0%
Supplier: TCI America
B0769-25ML
B0769-500ML
B0769-100ML
TCB0769-025MLEA
43.37
CAD
TCB0769-025ML
TCB0769-500ML
TCB0769-100ML
Butyrophenone ≥99.0%
Butyrophenone
CAS Number: 495-40-9
MDL Number: MFCD00009397
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 221
Melting point (°C): 12
Flash Point (°C): 88
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: Phenyl Propyl Ketone
Formula:
C₆H₅C(O)(CH₂)₂CH₃ Boiling Pt: 221 °C Melting Pt: 12 °C Density: 0.99 (20 °C) Flash Pt: 88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009397 CAS Number: 495-40-9 EINECS: 207-799-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...