Benzyl butyrate ≥97.0%

Supplier: TCI America

B0756-25G B0756-500G
TCB0756-025GEA 26.86 CAD
TCB0756-025G TCB0756-500G
Benzyl butyrate ≥97.0%
Benzyl butyrate

CAS Number: 103-37-7
MDL Number: MFCD00027133
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 114
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: Butyric Acid Benzyl Ester

Formula: C₁₁H₁₄O₂
Boiling Pt: 240 °C
Density: 1.01 (20 °C)
Flash Pt: 114 °C
MDL Number: MFCD00027133
CAS Number: 103-37-7
EINECS: 203-105-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45cf5d82cdb966bedbd88693bffea14c89f1089f&vid=45cf5d82cdb966bedbd88693bffea14c89f1089f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR