4-tert-Butylthiophenol ≥97.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-tert-Butyl-4-mercaptobenzene, 4-tert-Butylbenzene-1-thiol, 4-tert-Butylbenzenethiol
B0741-5G
B0741-25G
TCB0741-005GEA
104.28
CAD
TCB0741-005G
TCB0741-025G
4-tert-Butylthiophenol ≥97.0%
4-tert-Butylthiophenol
CAS Number: 2396-68-1
MDL Number: MFCD00022067
Molecular Formula: C10H14S
Molecular Weight: 166.28
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 0.99
Other Product Names / Synonyms: 4-tert-Butylthiophenol
Formula:
C₁₀H₁₄S MW: 166.29 g/mol Boiling Pt: 120 °C Melting Pt: –11 °C Density: 0.99 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00022067 CAS Number: 2396-68-1 EINECS: 219-255-6 UN: 3334 ADR: 9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...