Dibutyl sulfide ≥98.0%

Supplier: TCI America

B0740-500ML B0740-25ML
TCB0740-500MLEA 282.82 CAD
TCB0740-500ML TCB0740-025ML
Dibutyl sulfide ≥98.0%
Dibutyl sulphide

CAS Number: 544-40-1
MDL Number: MFCD00009468
Molecular Formula: C8H18S
Molecular Weight: 146.29
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Flash Point (°C): 76
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: Dibutyl Sulfide

Formula: C₈H₁₈S
Boiling Pt: 182 °C
Melting Pt: –80 °C
Density: 0.84 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00009468
CAS Number: 544-40-1
EINECS: 208-870-5
UN: 3334
ADR: 9,
Merck Index: 12,01626
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=192c9a91fb2dda4423d6e8e29325975b39103e85&vid=192c9a91fb2dda4423d6e8e29325975b39103e85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR