1-Butanol ≥99.0%
Supplier: TCI America
Synonyms:
n-Butanol
, Propyl carbinol
, 1-Butyl alcohol
, Butyl Alcohol
, 1-Hydroxy butane
, n-Butyl Alcohol
CAS Number: 71-36-3
MDL Number: MFCD00002964
Molecular Formula: C4H10O
Molecular Weight: 74.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Melting point (°C): -90
Flash Point (°C): 37
Specific Gravity (20/20): 0.81
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Butyl Alcohol
Formula:
H₃C(CH₂)₂CH₂OH MW: 74.12 g/mol Boiling Pt: 118 °C Melting Pt: –90 °C Density: 0.81 (20 °C) Flash Pt: 37 °C |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 UN: 1120 ADR: 3,III Merck Index: 13,01539 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...