2-Butoxyethyl acetate ≥98.0%

Supplier: TCI America
Warning

B0700-25ML B0700-500ML
TCB0700-25MLEA 36.34 CAD
TCB0700-25ML TCB0700-500ML
2-Butoxyethyl acetate ≥98.0%
2-Butoxyethyl acetate

CAS Number: 112-07-2
MDL Number: MFCD00009457
Molecular Formula: C8H16O3
Molecular Weight: 160.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 192
Melting point (°C): -64
Flash Point (°C): 71
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: Acetic Acid 2-Butoxyethyl Ester, 2-Butoxyethyl Acetate, Butyl Cellosolve Acetate, Butyl Glycol Acetate

Formula: C₈H₁₆O₃
Boiling Pt: 192 °C
Melting Pt: –64 °C
Density: 0.94 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00009457
CAS Number: 112-07-2
EINECS: 203-933-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e98a864dc2033be588e6f4790376c4b4a8816af&vid=0e98a864dc2033be588e6f4790376c4b4a8816af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR