Diethylene glycol monobutyl ether ≥99.0%

Supplier: TCI America
Warning

B0699-25ML B0699-500ML
TCB0699-25MLEA 33.29 CAD
TCB0699-25ML TCB0699-500ML
Diethylene glycol monobutyl ether ≥99.0%
Diethylene glycol monobutyl ether

CAS Number: 112-34-5
MDL Number: MFCD00002881
Molecular Formula: C8H18O3
Molecular Weight: 162.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Melting point (°C): -68
Flash Point (°C): 78
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 2-(2-Butoxyethoxy)ethanol, Butyl Carbitol

Formula: CH₃(CH₂)₃OCH₂CH₂OCH₂CH₂OH
Boiling Pt: 230 °C
Melting Pt: –68 °C
Density: 0.96 (20 °C)
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00002881
CAS Number: 112-34-5
EINECS: 203-961-6
Merck Index: 13,01556
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15a9966febfa63279489a2ebf443ca9ea3f5ad61&vid=15a9966febfa63279489a2ebf443ca9ea3f5ad61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR