cis-2-Butene-1,4-diol ≥94.0%

Supplier: TCI America
Warning

Synonyms: (Z)-But-2-ene-1,4-diol, (Z)-2-Butene-1,4-diol

B0693-500G B0693-25G
TCB0693-500GEA 111.55 CAD
TCB0693-500G TCB0693-025G
cis-2-Butene-1,4-diol ≥94.0%
cis-2-Butene-1,4-diol

CAS Number: 6117-80-2
MDL Number: MFCD00002924
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Melting point (°C): 7
Flash Point (°C): 128
Specific Gravity (20/20): 1.08


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: cis-1,4-Dihydroxy-2-butene

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 235
Melting Pt: 7 °C
Density: 1.08 (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002924
CAS Number: 6117-80-2
EINECS: 228-085-1
Merck Index: 12,02390
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c83432befd8301982f77fb0ed1e759cc7c70ac2b&vid=c83432befd8301982f77fb0ed1e759cc7c70ac2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR