2,3-Butanedione monoxime ≥98.0%

Supplier: TCI America

B0683-25G B0683-100G
TCB0683-025GEA 114.08 CAD
TCB0683-025G TCB0683-100G
2,3-Butanedione monoxime ≥98.0%
2,3-Butanedione monoxime

CAS Number: 57-71-6
MDL Number: MFCD00002116
Molecular Formula: C4H7NO2
Molecular Weight: 101.11
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 186
Melting point (°C): 76


Product Categories: Bioscience, Inhibitors (for Pharmacological Research)
Other Product Names / Synonyms: Biacetyl Monoxime, 2,3-Butanedione 2-Oxime, Isonitrosoethyl Methyl Ketone

Formula: C₄H₇NO₂
Boiling Pt: 186 °C
Melting Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00002116
CAS Number: 57-71-6
EINECS: 200-348-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8896cf6041c12b2ae6b9c2d2c5c70eec77783d7d&vid=8896cf6041c12b2ae6b9c2d2c5c70eec77783d7d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR