(±)-1,3-Butanediol ≥99.0%

Supplier: TCI America

B0679-500G B0679-25G
TCB0679-500GEA 61.78 CAD
TCB0679-500G TCB0679-025G
(±)-1,3-Butanediol ≥99.0%
(±)-1,3-Butanediol

CAS Number: 107-88-0
MDL Number: MFCD00004554
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 207
Flash Point (°C): 122
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 1,3-Butylene Glycol, 1,3-Dihydroxybutane

Formula: C₄H₁₀O₂
Boiling Pt: 207 °C
Melting Pt: < –50 °C
Density: 1.01 (20 °C)
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00004554
CAS Number: 107-88-0
EINECS: 203-529-7
Merck Index: 13,01566
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1390368ce7497f4408a93d745ba3eed2652d26ca&vid=781bf8b3058f0b3b3317afcf214a9067502d3772&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR