2-Bromo-m-xylene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Bromo-1,3-dimethylbenzene, 2,6-Dimethylbromobenzene

B0669-25G B0669-250G
TCB0669-025GEA 126.4 CAD
TCB0669-025G TCB0669-250G
2-Bromo-m-xylene ≥98.0%
2-Bromo-m-xylene

CAS Number: 576-22-7
MDL Number: MFCD00000075
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Melting point (°C): -10
Flash Point (°C): 74
Specific Gravity (20/20): 1.38


Other Product Names / Synonyms: 2,6-Dimethylbromobenzene

Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 206 °C
Melting Pt: –10 °C
Density: 1.38 (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00000075
CAS Number: 576-22-7
EINECS: 209-397-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1e46eabc8bee43ccd2b6f6ddd645c01efc52cdb&vid=05ba0f96106cbb2164a8810156abf5895a06ba80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR