5-Bromouridine ≥99.0% (by HPLC)

Supplier: TCI America

B0666-1G B0666-100MG
TCB0666-1GEA 135.88 CAD
TCB0666-1G TCB0666-100MG
5-Bromouridine ≥99.0% (by HPLC)
5-Bromouridine

CAS Number: 957-75-5
MDL Number: MFCD00006528
Molecular Formula: C9H11BrN2O6
Molecular Weight: 323.10
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 182
Specific rotation [a]20/D: -12 deg (C=2, H2O)
Storage Temperature: 0-10°C

Formula: C₉H₁₁BrN₂O₆
Melting Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00006528
CAS Number: 957-75-5
EINECS: 213-486-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea99c00116ef91de710745cb0fdf5b7b97633dae&vid=ea99c00116ef91de710745cb0fdf5b7b97633dae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR