2-Bromotoluene ≥98.0%
Supplier: TCI America
B0659-500G
B0659-25G
TCB0659-500GEA
334.96
CAD
TCB0659-500G
TCB0659-025G
2-Bromotoluene ≥98.0%
2-Bromotoluene
CAS Number: 95-46-5
MDL Number: MFCD00000068
Molecular Formula: C7H7Br
Molecular Weight: 171.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 181
Melting point (°C): -26
Flash Point (°C): 79
Specific Gravity (20/20): 1.42
Formula:
C₇H₇Br Boiling Pt: 181 °C Melting Pt: –26 °C Density: 1.42 (20 °C) Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000068 CAS Number: 95-46-5 EINECS: 202-421-7 UN: 3082 ADR: 9,III Merck Index: 13,01424 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...