Bromothymol blue 0.04% in water
Supplier: TCI America
Synonyms:
3,3-Bis(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
, 3′,3″-Dibromothymolsulfonphthalein
, 3′,3″-Dibromothymolsulphonphthalein
B0658-500ML
TCB0658-500MLEA
143.81
CAD
TCB0658-500ML
Bromothymol blue 0.04% in water
Bromothymol blue
[for pH Determination]
CAS Number: 76-59-5
MDL Number: MFCD00005872
Molecular Weight: 624.38
Form: Clear Liquid
Color: Brown
Formula:
C₂₇H₂₈Br₂O₅S MW: 624.39 g/mol |
MDL Number:
MFCD00005872 CAS Number: 76-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...