Bromothymol blue

Supplier: TCI America

B0657-1G B0657-25G
TCB0657-001GEA 68.24 CAD
TCB0657-001G TCB0657-025G
Bromothymol blue
Bromothymol blue

CAS Number: 76-59-5
MDL Number: MFCD00005872
Molecular Formula: C27H28Br2O5S
Molecular Weight: 624.38
Form: Crystal
Color: Pale Red
Melting point (°C): 202
Lambda max.: 615 nm (pH 8.0 buffer sol.)

Formula: C₂₇H₂₈Br₂O₅S
Melting Pt: 202 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e008f533cea36397aba217ea6b3fcaa9aea0fe0&vid=3e008f533cea36397aba217ea6b3fcaa9aea0fe0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR