Methyl-3-bromopropionate ≥99.0%

Supplier: TCI America

Synonyms: Methyl 3-bromopropanoate

B0648-25G B0648-100G
TCB0648-500GEA 712.04 CAD
TCB0648-500G TCB0648-025G TCB0648-100G
Methyl-3-bromopropionate ≥99.0%
Methyl 3-bromopropionate

CAS Number: 3395-91-3
MDL Number: MFCD00000250
Molecular Formula: C4H7BrO2
Molecular Weight: 167.00
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): 75
Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: 3-Bromopropionic Acid Methyl Ester

Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 105 °C
Density: 1.53 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00000250
CAS Number: 3395-91-3
EINECS: 222-247-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fe9e721219d8d7e27f94e61a5dcf1392bae6265&vid=5fe9e721219d8d7e27f94e61a5dcf1392bae6265&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR