3-Bromo-1-propanol ≥93.0%

Supplier: TCI America
Warning

Synonyms: 3-Bromopropan-1-ol, 1-Bromo-3-hydroxypropane

B0641-25G B0641-250G
TCB0641-025GEA 184.86 CAD
TCB0641-025G TCB0641-250G
3-Bromo-1-propanol ≥93.0%
3-Bromo-1-propanol

CAS Number: 627-18-9
MDL Number: MFCD00002942
Molecular Formula: C3H7BrO
Molecular Weight: 138.99
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 102
Specific Gravity (20/20): 1.60


Other Product Names / Synonyms: Trimethylene Bromohydrin

Formula: BrH₂CCH₂CH₂OH
MW: 138.99 g/mol
Boiling Pt: 140 °C
Density: 1.60 (20 °C)
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002942
CAS Number: 627-18-9
EINECS: 210-986-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7de71ecffe92a2b11065429627b4248fc1fa607d&vid=7de71ecffe92a2b11065429627b4248fc1fa607d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR