Bromophenol blue

Supplier: TCI America

B0631-1G B0631-25G
TCB0631-001GEA 33.18 CAD
TCB0631-001G TCB0631-025G
Bromophenol blue
Bromophenol blue

CAS Number: 115-39-9
MDL Number: MFCD00005875
Molecular Formula: C19H10Br4O5S
Molecular Weight: 669.96
Form: Crystal
Color: Pale Yellow
Melting point (°C): 273

Formula: C₁₉H₁₀Br₄O₅S
Melting Pt: 273 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44d256aaca209dd713803b55e3399449033a290c&vid=44d256aaca209dd713803b55e3399449033a290c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR