3-Bromophenol ≥95.0%

Supplier: TCI America

B0629-25G B0629-500G B0629-100G
TCB0629-025GEA 75.4 CAD
TCB0629-025G TCB0629-500G TCB0629-100G
3-Bromophenol ≥95.0%
3-Bromophenol

CAS Number: 591-20-8
MDL Number: MFCD00002253
Molecular Formula: C6H5BrO
Molecular Weight: 173.01
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 237
Flash Point (°C): 110
Freezing point (°C): 30

Formula: C₆H₅BrO
Boiling Pt: 237 °C
Melting Pt: 27…30 °C
Density: 0.641 g/cm³ (20 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002253
CAS Number: 591-20-8
EINECS: 209-706-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,01413
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49e1f46d845f93dbbf509dfd09f26b441a05967a&vid=e27c6fecbbca949478cbe8832a8ff8c38d40755d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR