6-Bromo-2-naphthol ≥98.0%

Supplier: TCI America

Synonyms: 6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene

B0621-25G B0621-100G
TCB0621-025GEA 62.55 CAD
TCB0621-025G TCB0621-100G
6-Bromo-2-naphthol ≥98.0%
6-Bromo-2-naphthol

CAS Number: 15231-91-1
MDL Number: MFCD00004081
Molecular Formula: C10H7BrO
Molecular Weight: 223.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 129

Formula: C₁₀H₇BrO
MW: 223.07 g/mol
Boiling Pt: 200…205 °C (20 mmHg)
Melting Pt: 129 °C
MDL Number: MFCD00004081
CAS Number: 15231-91-1
EINECS: 239-279-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9285e654966b375d9035cbe0dd5b7a455434e8e&vid=b9285e654966b375d9035cbe0dd5b7a455434e8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR