2-Bromonaphthalene ≥98.0%

Supplier: TCI America
Warning

B0619-25G B0619-5G
TCB0619-025GEA 139.04 CAD
TCB0619-025G TCB0619-005G
2-Bromonaphthalene ≥98.0%
2-Bromonaphthalene

CAS Number: 580-13-2
MDL Number: MFCD00004051
Molecular Formula: C10H7Br
Molecular Weight: 207.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 282
Melting point (°C): 58
Flash Point (°C): 110

Formula: C₁₀H₇Br
MW: 207.07 g/mol
Boiling Pt: 282 °C
Melting Pt: 58 °C
Density: 1.605 g/cm³ (0 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004051
CAS Number: 580-13-2
EINECS: 209-452-5
Merck Index: 13,01411
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06c9cea839aed4cac329504b3fc5370327770cdf&vid=06c9cea839aed4cac329504b3fc5370327770cdf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR