1-Bromo-4-iodobenzene ≥98.0%
Supplier: TCI America
|
Warning
|
B0604-250G
B0604-25G
TCB0604-250GEA
673.08
CAD
TCB0604-250G
TCB0604-025G
1-Bromo-4-iodobenzene ≥98.0%
1-Bromo-4-iodobenzene
CAS Number: 589-87-7
MDL Number: MFCD00001051
Molecular Formula: C6H4BrI
Molecular Weight: 282.91
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 122
Melting point (°C): 91
Other Product Names / Synonyms: 4-Bromoiodobenzene
Formula:
C₆H₄BrI Boiling Pt: 122 °C Melting Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001051 CAS Number: 589-87-7 EINECS: 209-662-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...