N-(2-Bromoethyl)phthalimide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2-Bromoethyl)isoindoline-1,3-dione

B0597-25G B0597-100G
TCB0597-025GEA 77.42 CAD
TCB0597-025G TCB0597-100G
N-(2-Bromoethyl)phthalimide ≥98.0% (by GC, titration analysis)
N-(2-Bromoethyl)phthalimide

CAS Number: 574-98-1
MDL Number: MFCD00005902
Molecular Formula: C10H8BrNO2
Molecular Weight: 254.08
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 318
Melting point (°C): 82

Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Boiling Pt: 318 °C
Melting Pt: 82 °C
MDL Number: MFCD00005902
CAS Number: 574-98-1
EINECS: 209-379-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7142673f15e74bc6496cd514f10cc01b5033565d&vid=7142673f15e74bc6496cd514f10cc01b5033565d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR