2-Ethylhexyl bromide ≥97.0%

Supplier: TCI America

B0596-25ML B0596-500ML B0596-100ML
TCB0596-025MLEA 26.86 CAD
TCB0596-025ML TCB0596-500ML TCB0596-100ML
2-Ethylhexyl bromide ≥97.0%
2-Ethylhexyl bromide

CAS Number: 18908-66-2
MDL Number: MFCD00000220
Molecular Formula: C8H17Br
Molecular Weight: 193.13
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 156
Flash Point (°C): 69
Specific Gravity (20/20): 1.12


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 3-(Bromomethyl)heptane, 2-Ethylhexyl Bromide, Octyl Bromide

Formula: C₈H₁₇Br
Boiling Pt: 156 °C
Density: 1.12 (20 °C)
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000220
CAS Number: 18908-66-2
EINECS: 242-659-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78cb7c7e43da79c97ff5dff88b54c24e29358a40&vid=11b765d859db9e5f9d76f41fa0614c2287f001fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR