Phenethyl bromide ≥98.0%

Supplier: TCI America
Warning

Synonyms: (2-Bromoethyl)benzene, 2-Phenylethyl bromide

B0594-25G B0594-500G B0594-100G
TCB0594-025GEA 28.28 CAD
TCB0594-025G TCB0594-500G TCB0594-100G
Phenethyl bromide ≥98.0%
Phenethyl bromide

CAS Number: 103-63-9
MDL Number: MFCD00000240
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 96
Specific Gravity (20/20): 1.37
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Phenethyl Bromide, 2-Phenylethyl Bromide

Formula: BrCH₂CH₂C₆H₅
Boiling Pt: 220 °C
Melting Pt: –56 °C
Density: 1.37 (20 °C)
Flash Pt: 96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000240
CAS Number: 103-63-9
EINECS: 203-130-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f1e02460a0ad524b7db76d2c91b1000373c8668&vid=77f0dd7593c12fc92c78f84aa49717d00d1600a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR