(1-Bromoethyl)benzene ≥95.0%
Supplier: TCI America
Synonyms:
1-Bromo-1-phenylethane, α-Methylbenzyl bromide
B0592-25G
B0592-100G
TCB0592-025GEA
64.59
CAD
TCB0592-025G
TCB0592-100G
(1-Bromoethyl)benzene ≥95.0%
1-Bromo-1-phenylethane
CAS Number: 585-71-7
MDL Number: MFCD00000139
Molecular Formula: C8H9Br
Molecular Weight: 185.06
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 96
Specific Gravity (20/20): 1.36
Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: 1-Phenylethyl Bromide
Formula:
C₆H₅CHBrCH₃ Boiling Pt: 200 °C Melting Pt: 50 °C Density: 1.36 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00000139 CAS Number: 585-71-7 EINECS: 209-560-2 UN: 3265 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...