2-Bromethylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

B0591-25G B0591-500G B0591-100G
TCB0591-025GEA 67.94 CAD
TCB0591-025G TCB0591-500G TCB0591-100G
2-Bromethylamine hydrobromide ≥98.0% (by total nitrogen and titration analysis)
2-Bromethylamine hydrobromide

CAS Number: 2576-47-8
MDL Number: MFCD00012886
Molecular Formula: C2H6BrN
Molecular Weight: 204.89
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 175


Other Product Names / Synonyms: 2-Aminoethyl Bromide Hydrobromide

Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
EINECS: 219-924-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=649f03a9cf7c4bff37af44e912d6021a536574de&vid=649f03a9cf7c4bff37af44e912d6021a536574de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR