1-Bromododecane ≥98.0%
Supplier: TCI America
B0587-25ML
B0587-500ML
TCB0587-025MLEA
49.63
CAD
TCB0587-025ML
TCB0587-500ML
1-Bromododecane ≥98.0%
1-Bromododecane
CAS Number: 143-15-7
MDL Number: MFCD00000225
Molecular Formula: C12H25Br
Molecular Weight: 249.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 146
Specific Gravity (20/20): 1.04
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Dodecyl Bromide, Lauryl Bromide
Formula:
BrH₂C(CH₂)₁₀CH₃ Boiling Pt: 180 °C Melting Pt: –10 °C Density: 1.04 (20 °C) Flash Pt: 146 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000225 CAS Number: 143-15-7 EINECS: 205-587-9 Merck Index: 13,05406 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...