4-Bromo-N,N-dimethylaniline ≥98.0%

Supplier: TCI America

B0585-25G B0585-500G B0585-100G
TCB0585-025GEA 56.25 CAD
TCB0585-025G TCB0585-500G TCB0585-100G
4-Bromo-N,N-dimethylaniline ≥98.0%
4-Bromo-N,N-dimethylaniline

CAS Number: 586-77-6
MDL Number: MFCD00000093
Molecular Formula: C8H10BrN
Molecular Weight: 200.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 264
Melting point (°C): 54
Flash Point (°C): 110

Formula: C₈H₁₀BrN
Boiling Pt: 264 °C
Melting Pt: 54 °C
Density: 1.3218 g/cm³ (99.5 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000093
CAS Number: 586-77-6
EINECS: 209-582-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1e8c603d5e9012be67435c46712a6ac8040935c&vid=e1e8c603d5e9012be67435c46712a6ac8040935c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR