1-Bromodecane ≥98.0%

Supplier: TCI America
Warning

B0583-500ML B0583-25ML
TCB0583-500MLEA 84.72 CAD
TCB0583-500ML TCB0583-025ML
1-Bromodecane ≥98.0%
1-Bromodecane

CAS Number: 112-29-8
MDL Number: MFCD00000221
Molecular Formula: C10H21Br
Molecular Weight: 221.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Melting point (°C): -30
Flash Point (°C): 94
Specific Gravity (20/20): 1.07


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Decyl Bromide

Formula: BrH₂C(CH₂)₈CH₃
Boiling Pt: 139 °C
Melting Pt: –30 °C
Density: 1.07 (20 °C)
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000221
CAS Number: 112-29-8
EINECS: 203-955-3
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5a9d0936f7ac6adeb3c30f6084d1c154c4f0278&vid=0ec8db18f71d397381da6f19341419d0b7db1317&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR