Bromocyclohexane ≥98.0%

Supplier: TCI America

B0581-25G B0581-500G B0581-100G
TCB0581-025GEA 46.64 CAD
TCB0581-025G TCB0581-500G TCB0581-100G
Bromocyclohexane ≥98.0%
Bromocyclohexane

CAS Number: 108-85-0
MDL Number: MFCD00003819
Molecular Formula: C6H11Br
Molecular Weight: 163.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 62
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: Cyclohexyl Bromide

Formula: C₆H₁₁Br
Boiling Pt: 165 °C
Melting Pt: –56.5 °C
Density: 1.33 (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003819
CAS Number: 108-85-0
EINECS: 203-622-2
Merck Index: 13,02759
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3516f7490d7c361c5d28fd36b5f622fdbd6da4e8&vid=6e7af25620e6c885c442c3707321586fcd67ec84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR