Bromocresol purple ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 5',5''-Dibromo-o-cresolsulfonphthalein

B0580-1G B0580-25G
TCB0580-001GEA 59.49 CAD
TCB0580-001G TCB0580-025G
Bromocresol purple ≥97.0% (by HPLC)
Bromocresol purple

CAS Number: 115-40-2
MDL Number: MFCD00011681
Molecular Formula: C21H16Br2O5S
Molecular Weight: 540.22
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 240

Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
Melting Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c900df847e0adb7b4e44259555455f37b474eec7&vid=c900df847e0adb7b4e44259555455f37b474eec7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR