Bromocresol green

Supplier: TCI America

B0578-25G B0578-1G B0578-5G
TCB0578-025GEA 309.68 CAD
TCB0578-025G TCB0578-001G TCB0578-5G
Bromocresol green
Bromocresol green

CAS Number: 76-60-8
MDL Number: MFCD00005874
Molecular Formula: C21H14Br4O5S
Molecular Weight: 698.01
Form: Crystal
Color: White
Melting point (°C): 219


Other Product Names / Synonyms: Bromocresol Blue

Formula: C₂₁H₁₄Br₄O₅S
Melting Pt: 219 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4aa78542c26fccce426ef742c29cc0e78600792&vid=d4aa78542c26fccce426ef742c29cc0e78600792&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR