1-Bromo-4-chlorobenzene ≥99.0%

Supplier: TCI America

Synonyms: 4-Bromochlorobenzene

B0571-25G B0571-500G B0571-100G
TCB0571-025GEA 30.56 CAD
TCB0571-025G TCB0571-500G TCB0571-100G
1-Bromo-4-chlorobenzene ≥99.0%
1-Bromo-4-chlorobenzene

CAS Number: 106-39-8
MDL Number: MFCD00000600
Molecular Formula: C6H4BrCl
Molecular Weight: 191.45
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 196
Melting point (°C): 67
Flash Point (°C): 61


Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 4-Bromochlorobenzene

Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C
Melting Pt: 67 °C
Density: 1.576 g/cm³ (71 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8df08256b77ce27db9eb1d056c3f884af1b843c2&vid=c5c210fefaf3b78bce3feeec14286f01fab55475&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR