1-Bromo-3-chlorobenzene ≥99.0%
Supplier: TCI America
|
Warning
|
B0570-25G
B0570-100G
TCB0570-025GEA
48.98
CAD
TCB0570-025G
TCB0570-100G
1-Bromo-3-chlorobenzene ≥99.0%
1-Bromo-3-chlorobenzene
CAS Number: 108-37-2
MDL Number: MFCD00000568
Molecular Formula: C6H4BrCl
Molecular Weight: 191.45
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 80
Specific Gravity (20/20): 1.63
Product Categories: Synthetic Organic Chemistry, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 3-Bromochlorobenzene
Formula:
C₆H₄BrCl Boiling Pt: 196 °C Melting Pt: –21.2 °C Density: 1.63 (20 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000568 CAS Number: 108-37-2 EINECS: 203-575-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...