1-Bromo-2-chlorobenzene ≥99.0%
Supplier: TCI America
|
Warning
|
B0569-25G
B0569-500G
TCB0569-025GEA
42.66
CAD
TCB0569-025G
TCB0569-500G
1-Bromo-2-chlorobenzene ≥99.0%
1-Bromo-2-chlorobenzene
CAS Number: 694-80-4
MDL Number: MFCD00000532
Molecular Formula: C6H4BrCl
Molecular Weight: 191.45
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 204
Flash Point (°C): 79
Specific Gravity (20/20): 1.65
Other Product Names / Synonyms: 2-Bromochlorobenzene
Formula:
C₆H₄BrCl Boiling Pt: 204 °C Melting Pt: –12.1 °C Density: 1.65 (20 °C) Flash Pt: 79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000532 CAS Number: 694-80-4 EINECS: 211-775-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...