Methyl-2-bromobenzoate ≥98.0%

Supplier: TCI America

B0555-250G B0555-25G
TCB0555-250GEA 380.78 CAD
TCB0555-250G TCB0555-25G
Methyl-2-bromobenzoate ≥98.0%
Methyl 2-bromobenzoate

CAS Number: 610-94-6
MDL Number: MFCD00016328
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 253
Flash Point (°C): 113
Specific Gravity (20/20): 1.53


Other Product Names / Synonyms: 2-Bromobenzoic Acid Methyl Ester

Formula: C₈H₇BrO₂
Boiling Pt: 253 °C
Density: 1.53 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00016328
CAS Number: 610-94-6
EINECS: 210-241-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a64696601f0cbdc4eb54affdc4ff08115080370&vid=0a64696601f0cbdc4eb54affdc4ff08115080370&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR