Methyl-3-bromobenzoate ≥99.0%

Supplier: TCI America

B0554-5G B0554-25G
TCB0554-5GEA 53.72 CAD
TCB0554-5G TCB0554-025G
Methyl-3-bromobenzoate ≥99.0%
Methyl 3-bromobenzoate

CAS Number: 618-89-3
MDL Number: MFCD00017777
Molecular Formula: C8H7BrO2
Molecular Weight: 215.05
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 30


Other Product Names / Synonyms: 3-Bromobenzoic Acid Methyl Ester

Formula: C₈H₇BrO₂
Boiling Pt: 69…70 °C (0.2 mmHg)
Melting Pt: 31 °C
Flash Pt: 110 °C
MDL Number: MFCD00017777
CAS Number: 618-89-3
EINECS: 210-569-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4e9a914b46b24409ff451b9a5ef964edd060162&vid=c4e9a914b46b24409ff451b9a5ef964edd060162&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR