4-Bromobenzoic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: p-Bromobenzoic Acid

B0553-25G B0553-100G
TCB0553-500GEA 599.52 CAD
TCB0553-500G TCB0553-025G TCB0553-100G
4-Bromobenzoic acid ≥98.0%
4-Bromobenzoic acid

CAS Number: 586-76-5
MDL Number: MFCD00002529
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 255

Formula: C₇H₅BrO₂
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 255 °C
Density: 1.894 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7
Merck Index: 13,01392
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f77d7df4d690c85de033cbec66a327e27a39189&vid=0ffa27dd36259b38c6e9833814ab956fceb4f815&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR