2-Bromobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: o-Bromobenzoic Acid

B0552-25G B0552-500G B0552-100G
TCB0552-025GEA 30.02 CAD
TCB0552-025G TCB0552-500G TCB0552-100G
2-Bromobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromobenzoic acid

CAS Number: 88-65-3
MDL Number: MFCD00002402
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 150
Flash Point (°C): 100

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 150 °C
Density: 1.929 g/cm³ (20 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3
EINECS: 201-848-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=261b397d74a937da7624890b93c154e778149319&vid=f379a8d456c3d1757515efa0c721a34fd89ae09b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR