3-Bromobenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
m-Bromobenzoic Acid
B0551-25G
B0551-250G
TCB0551-025GEA
91.64
CAD
TCB0551-025G
TCB0551-250G
3-Bromobenzoic acid ≥98.0%
3-Bromobenzoic acid
CAS Number: 585-76-2
MDL Number: MFCD00002487
Molecular Formula: C7H5BrO2
Molecular Weight: 201.02
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 158
Formula:
C₇H₅BrO₂ Boiling Pt: 308 °C (1013 hPa) Melting Pt: 158 °C Density: 1.845 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002487 CAS Number: 585-76-2 EINECS: 209-562-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...